Learn More
Medchemexpress LLC Guanabenz hydrochloride | 23113-43-1 | 99.9% | 100 MG
SDP
Catalog No. 5000340046
Encompass_Preferred
Guanabenz hydrochloride is an orally active α-2-adrenoceptor agonist. It exhibits antihypertensive and antiparasitic activity, interferes with ER stress-signalling, and offers protective effects in cardiac myocytes. It is also used for research related to high blood pressure.
  • Orally active α-2-adrenoceptor agonist
  • Exhibits antihypertensive activity
  • Possesses antiparasitic activity
  • Interferes with ER stress-signaling
  • Offers protective effects in cardiac myocytes
  • Used for research related to high blood pressure
Catalog No. 50-003-40046 Supplier Medchemexpress LLC Supplier No. HY12724A100MG
Guanabenz hydrochloride is an orally active α-2-adrenoceptor agonist. It exhibits antihypertensive and antiparasitic activity, interferes with ER stress-signalling, and offers protective effects in cardiac myocytes. It is also used for research related to high blood pressure.
  • Orally active α-2-adrenoceptor agonist
  • Exhibits antihypertensive activity
  • Possesses antiparasitic activity
  • Interferes with ER stress-signaling
  • Offers protective effects in cardiac myocytes
  • Used for research related to high blood pressure