Learn More
Medchemexpress LLC Guanabenz hydrochloride | 23113-43-1 | 99.9% | 25 MG
SDP
Catalog No. 5000338686
Encompass_Preferred
Guanabenz hydrochloride is an orally active α-2-adrenoceptor agonist with antihypertensive and antiparasitic activity. It interferes with ER stress-signalling and has protective effects in cardiac myocytes. It is also used for the research of high blood pressure. For research use only.
  • Orally active alpha-2-adrenoceptor agonist
  • Exhibits antihypertensive activity
  • Displays antiparasitic activity
  • Interferes with ER stress-signalling
  • Has protective effects in cardiac myocytes
  • Used for research of high blood pressure
Catalog No. 50-003-38686 Supplier Medchemexpress LLC Supplier No. 25MGHY12724A
Guanabenz hydrochloride is an orally active α-2-adrenoceptor agonist with antihypertensive and antiparasitic activity. It interferes with ER stress-signalling and has protective effects in cardiac myocytes. It is also used for the research of high blood pressure. For research use only.
  • Orally active alpha-2-adrenoceptor agonist
  • Exhibits antihypertensive activity
  • Displays antiparasitic activity
  • Interferes with ER stress-signalling
  • Has protective effects in cardiac myocytes
  • Used for research of high blood pressure