Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanethidine sulfate | 645-43-2 | 98.0% | 1 ML
SDP

Catalog No. 5000408065
Encompass_Preferred

Guanethidine sulfate is an antihypertensive agent that functions as an adrenergic neurone blocking agent. It gains entry into noradrenergic nerve terminals via the neuronal amine carrier.

  • Functions as an antihypertensive agent
  • Acts as an adrenergic neurone blocking agent
  • Enters noradrenergic nerve terminals via the neuronal amine carrier
  • Has a purity of 98.0%
  • Solution stable for 6 months at -80°C
  • Solution stable for 1 month at -20°C

Catalog No. 50-004-08065 Supplier Medchemexpress LLC Supplier No. HYB080010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guanethidine sulfate is an antihypertensive agent that functions as an adrenergic neurone blocking agent. It gains entry into noradrenergic nerve terminals via the neuronal amine carrier.

  • Functions as an antihypertensive agent
  • Acts as an adrenergic neurone blocking agent
  • Enters noradrenergic nerve terminals via the neuronal amine carrier
  • Has a purity of 98.0%
  • Solution stable for 6 months at -80°C
  • Solution stable for 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.