Learn More
Medchemexpress LLC Guanethidine sulfate | 645-43-2 | 98.0% | 500 MG
SDP
Catalog No. 5000409324
Encompass_Preferred
Guanethidine sulfate is an antihypertensive and adrenergic neurone blocking agent. It functions by entering noradrenergic nerve terminals via the neuronal amine carrier. It has been studied for its effects in inducing sympathectomy and lowering blood pressure in animal models.
  • Antihypertensive agent
  • Adrenergic neurone blocking agent
  • Enters noradrenergic nerve terminals via neuronal amine carrier
  • Induces sympathectomy in animal models
  • Lowers blood pressure
  • Causes hypersensitivity to noradrenaline
Catalog No. 50-004-09324 Supplier Medchemexpress LLC Supplier No. HYB0800500MG
Guanethidine sulfate is an antihypertensive and adrenergic neurone blocking agent. It functions by entering noradrenergic nerve terminals via the neuronal amine carrier. It has been studied for its effects in inducing sympathectomy and lowering blood pressure in animal models.
  • Antihypertensive agent
  • Adrenergic neurone blocking agent
  • Enters noradrenergic nerve terminals via neuronal amine carrier
  • Induces sympathectomy in animal models
  • Lowers blood pressure
  • Causes hypersensitivity to noradrenaline