Learn More
Enzo Life Sciences Guanfacine. HCl (500mg). CAS: 29520-14-7 / 29110-48-3
SDP
Catalog No. 502010325
Encompass_Preferred
Centrally acting α2-adrenergic receptor agonist. In the guinea pig cerebral cortex, the Kd is 20 and 880 nM for the α2A- and the α2C-adrenoceptor, respectively. Antihypertensive agent. Purity: ≥98% (HPLC). Solubility: Soluble in water (25 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0325 Supplier Enzo Life Sciences Supplier No. BMLAR1020500
Centrally acting α2-adrenergic receptor agonist. In the guinea pig cerebral cortex, the Kd is 20 and 880 nM for the α2A- and the α2C-adrenoceptor, respectively. Antihypertensive agent. Purity: ≥98% (HPLC). Solubility: Soluble in water (25 mg/ml). Long Term Storage: Ambient.