Learn More
Medchemexpress LLC Guanfacine (hydrochloride) | 29110-48-3 | 100.0% | 282.55 | 100 MG
SDP
Catalog No. 5000385647
Encompass_Preferred
Guanfacine hydrochloride is an orally active and blood-brain barrier permeable noradrenergic α2A agonist, highly selective for the α2A receptor subtype. It has effects in producing hypotension and sedation, and can be used for research of various prefrontal cortex (PFC) cognitive disorders, including Tourette's syndrome and attention deficit hyperactivity disorder (ADHD).
  • Orally active and blood-brain barrier permeable
  • Highly selective α2A receptor subtype agonist
  • Produces hypotension and sedation
  • Used for research on prefrontal cortex cognitive disorders
  • Increases delay-related neuronal firing for working memory
  • Improves PFC cognitive function
  • Enhances PFC working memory in aged monkeys
  • Improves impulse control in delayed discounting tasks
  • Improves cognitive performance when infused into rat or monkey PFC
Catalog No. 50-003-85647 Supplier Medchemexpress LLC Supplier No. HY17416R100MG
Guanfacine hydrochloride is an orally active and blood-brain barrier permeable noradrenergic α2A agonist, highly selective for the α2A receptor subtype. It has effects in producing hypotension and sedation, and can be used for research of various prefrontal cortex (PFC) cognitive disorders, including Tourette's syndrome and attention deficit hyperactivity disorder (ADHD).
  • Orally active and blood-brain barrier permeable
  • Highly selective α2A receptor subtype agonist
  • Produces hypotension and sedation
  • Used for research on prefrontal cortex cognitive disorders
  • Increases delay-related neuronal firing for working memory
  • Improves PFC cognitive function
  • Enhances PFC working memory in aged monkeys
  • Improves impulse control in delayed discounting tasks
  • Improves cognitive performance when infused into rat or monkey PFC