Learn More
TARGETMOL CHEMICALS INC GUANFACINE HYDROCHLORIDE 50MG
SDP
Catalog No. 502577216
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Guanfacine hydrochloride (Intuniv) is a centrally acting antihypertensive agent with specificity towards ADRENERGIC ALPHA-2 RECEPTORS. purity: 99%
Catalog No. 50-257-7216 Supplier TARGETMOL CHEMICALS INC Supplier No. T215050MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Guanfacine hydrochloride (Intuniv) is a centrally acting antihypertensive agent with specificity towards ADRENERGIC ALPHA-2 RECEPTORS. purity: 99%