Learn More
Medchemexpress LLC Guangxitoxin 1E TFA | ≥95.0% | 3948.61 | 100 UG
SDP
Catalog No. 5000423903
Encompass_Preferred
Guangxitoxin 1E TFA is the trifluoroacetate salt form of Guangxitoxin 1E, a peptide toxin. It functions as a potent and selective blocker of voltage-gated potassium channels KV2.1 and KV2.2, with an IC50 of 1-3 nM. This compound is intended for research purposes only and is not for human therapeutic use.
  • Potent and selective blocker of KV2.1 and KV2.2 channels (IC50 of 1-3 nM)
  • Inhibits KV4.3 channels (IC50 of 24-54 nM)
  • Inhibits 90% of IDR in mouse β-cells
  • Shifts voltage dependence of channel activation to more depolarized potentials
  • Broadens β-cell action potential
  • Enhances glucose-stimulated intracellular calcium oscillations
  • Increases insulin secretion from mouse pancreatic islets in a glucose-dependent manner
Catalog No. 50-004-23903 Supplier Medchemexpress LLC Supplier No. HYP1427A100UG
Guangxitoxin 1E TFA is the trifluoroacetate salt form of Guangxitoxin 1E, a peptide toxin. It functions as a potent and selective blocker of voltage-gated potassium channels KV2.1 and KV2.2, with an IC50 of 1-3 nM. This compound is intended for research purposes only and is not for human therapeutic use.
  • Potent and selective blocker of KV2.1 and KV2.2 channels (IC50 of 1-3 nM)
  • Inhibits KV4.3 channels (IC50 of 24-54 nM)
  • Inhibits 90% of IDR in mouse β-cells
  • Shifts voltage dependence of channel activation to more depolarized potentials
  • Broadens β-cell action potential
  • Enhances glucose-stimulated intracellular calcium oscillations
  • Increases insulin secretion from mouse pancreatic islets in a glucose-dependent manner