Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Carbonate, MP Biomedicals

Catalog No. ICN209460 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN209460 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN209460 Supplier MP Biomedicals, Inc Supplier No. 0520946080

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3430-10-2
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
InChI Key ZSFPJJJRNUZCEV-UHFFFAOYSA-N
Synonym 3-amino-2-picoline, 3-amino-2-methylpyridine, 2-methyl-3-pyridinamine, 2-methyl-3-aminopyridine, 2-methyl-pyridin-3-ylamine, 3-pyridinamine, 2-methyl, 2-methyl-3-pyridinylamine, 2-methyl-3-pyridylamine, 2-methylpyridine-3-amine, zlchem 29
PubChem CID 572616
IUPAC Name 2-methylpyridin-3-amine
SMILES CC1=C(C=CC=N1)N

Specifications

Quantity 100 g
Formula Weight 180.17
Chemical Name or Material Guanidine Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.