Learn More
Medchemexpress LLC Guanidine-d5 hydrochloride | 108694-93-5 | 98% | 250 MG
SDP
Catalog No. 5000406823
Encompass_Preferred
Guanidine-d5 hydrochloride is the deuterium labeled Guanidine hydrochloride, a strong chaotrope and denaturant of proteins. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06823 Supplier Medchemexpress LLC Supplier No. 250MGHYB0178AS1
Guanidine-d5 hydrochloride is the deuterium labeled Guanidine hydrochloride, a strong chaotrope and denaturant of proteins. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS