Learn More
Medchemexpress LLC Guanidine-d5 hydrochloride | 108694-93-5 | MFCClD6N3 | 98% | 25 MG
SDP
Catalog No. 5000450598
Encompass_Preferred
Guanidine-d5 (hydrochloride) is the deuterium labeled Guanidine hydrochloride. Guanidine hydrochloride (Guanidinium chloride) is a strong chaotrope and also a strong denaturant of proteins.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-50598 Supplier Medchemexpress LLC Supplier No. 25MGHYB0178AS1
Guanidine-d5 (hydrochloride) is the deuterium labeled Guanidine hydrochloride. Guanidine hydrochloride (Guanidinium chloride) is a strong chaotrope and also a strong denaturant of proteins.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.