Learn More
Research Products International Corp Guanidine Hydrochloride, 100 Grams
SDP
Catalog No. 50213353
Encompass_Preferred
Protein denaturant used for purifying proteins and nucleic acids.
Catalog No. 50-213-353 Supplier Research Products International Corp Supplier No. G49000100.0
Add to Cart
Add to Cart
Protein denaturant used for purifying proteins and nucleic acids.