Learn More
Apexbio Technology LLC Guanidine HCl 50-01-1 50g
SDP
Catalog No. 5000094599
Encompass_Preferred
Guanidine hydrochloride (Guanidine HCl CAS 50-01-1) is a highly alkaline crystalline compound derived from the oxidative metabolism of guanine Functioning primarily as a strong chaotropic agent it disrupts hydrogen bonding and secondary structure in proteins leading to efficient protein denaturation and solubilization Widely utilized in molecular biology and biochemistry Guanidine HCl facilitates protein unfolding for downstream applications such as protein purification refolding studies and RNA extraction protocols where it also serves to inactivate nucleases and other enzymes
Catalog No. 50-000-94599 Supplier Apexbio Technology LLC Supplier No. B194950000
Guanidine hydrochloride (Guanidine HCl CAS 50-01-1) is a highly alkaline crystalline compound derived from the oxidative metabolism of guanine Functioning primarily as a strong chaotropic agent it disrupts hydrogen bonding and secondary structure in proteins leading to efficient protein denaturation and solubilization Widely utilized in molecular biology and biochemistry Guanidine HCl facilitates protein unfolding for downstream applications such as protein purification refolding studies and RNA extraction protocols where it also serves to inactivate nucleases and other enzymes