Learn More
Gold Biotechnology Inc Guanidine HCl 500 g
SDP
Catalog No. 5000562159
Encompass_Preferred
Guanidine HCl is a freely soluble chaotropic agent routinely used for protein denaturation and folding
Catalog No. 50-005-62159 Supplier Gold Biotechnology Inc Supplier No. G211500
Guanidine HCl is a freely soluble chaotropic agent routinely used for protein denaturation and folding