Learn More
Crescent Chemical Co Inc GUANIDINE-HCL MOL BIO GRADE
SDP
Catalog No. 50248387
Encompass_Preferred
Guanidine-HCl molecular biology grade DNase/RNase not detected. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 39558.02 CAS 50-01-1
Catalog No. 50-248-387 Supplier Crescent Chemical Co Inc Supplier No. SE39558.02
Guanidine-HCl molecular biology grade DNase/RNase not detected. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 39558.02 CAS 50-01-1