Learn More
Crescent Chemical Co Inc Guanidine-HCl, 50-01-1, MFCD00013026,l 1kg
SDP
Catalog No. 50247697
Encompass_Preferred
Molecular Formula CH5N3.ClH, Molecular Weight 95.532, Research Grade, Reagent for reversible denaturation and renaturation of proteins. Assay pH (6 M in water 20 °C) min. 99.0 %4.5 - 7.0. Storage Conditions: +15 °C TO +30 °C.
Catalog No. 50-247-697 Supplier Crescent Chemical Co Inc Supplier No. SE24205.02
Molecular Formula CH5N3.ClH, Molecular Weight 95.532, Research Grade, Reagent for reversible denaturation and renaturation of proteins. Assay pH (6 M in water 20 °C) min. 99.0 %4.5 - 7.0. Storage Conditions: +15 °C TO +30 °C.