Learn More
Alkali Scientific Guanidine Hydrochloride, 1 Kg
SDP
Catalog No. 5000659563
Encompass_Preferred
Guanidine Hydrochloride (1kg) is a high-purity reagent used in molecular biology for protein denaturation and nucleic acid isolation. This chaotropic agent is critical for unfolding proteins and disrupting their secondary, tertiary, and quaternary structures, making it essential for applications such as protein purification, enzyme studies, and nucleic acid extraction. It is particularly useful in preparing samples for electrophoresis and in isolating RNA and DNA from complex biological samples. Due to its strong denaturing properties, it also plays a role in the preparation of buffers for analytical and structural biology studies.
Catalog No. 50-006-59563 Supplier Alkali Scientific Supplier No. GUA002CP
Guanidine Hydrochloride (1kg) is a high-purity reagent used in molecular biology for protein denaturation and nucleic acid isolation. This chaotropic agent is critical for unfolding proteins and disrupting their secondary, tertiary, and quaternary structures, making it essential for applications such as protein purification, enzyme studies, and nucleic acid extraction. It is particularly useful in preparing samples for electrophoresis and in isolating RNA and DNA from complex biological samples. Due to its strong denaturing properties, it also plays a role in the preparation of buffers for analytical and structural biology studies.