Learn More
Alkali Scientific Guanidine Hydrochloride, 100 G
SDP
Supplier:  Alkali Scientific GUA001CP
Encompass
Guanidine Hydrochloride is a high-purity chaotropic agent commonly used in protein denaturation and nucleic acid isolation. This 100g sample is essential in disrupting protein structures for biochemical studies, facilitating the extraction of RNA, DNA, and proteins in molecular biology protocols. Its strong denaturing properties make it ideal for breaking down protein tertiary and quaternary structures, ensuring the efficient isolation of nucleic acids. It is also used in the preparation of buffers for experiments involving enzymatic activity and in studies of protein folding and structure. Guanidine Hydrochloride is indispensable in many molecular and biochemical laboratories.
Catalog No. 50-006-59646