Learn More
Alkali Scientific Guanidine Hydrochloride, 5 Kg
SDP
Catalog No. 5000659591
Encompass_Preferred
Guanidine Hydrochloride, available in a 5kg format, is a high-purity chaotropic agent used in the denaturation of proteins and isolation of nucleic acids. This large quantity is ideal for high-throughput molecular biology labs that require significant volumes of denaturing reagent for large-scale protein analysis, RNA, and DNA extraction. Its denaturing properties are critical for disrupting protein structures, making it a standard reagent in studies of protein folding, enzyme function, and nucleic acid isolation. Researchers use this compound in protein chemistry, structural biology, and high-resolution assays where precise denaturation is essential for achieving reliable results.
Catalog No. 50-006-59591 Supplier Alkali Scientific Supplier No. GUA004CP
Guanidine Hydrochloride, available in a 5kg format, is a high-purity chaotropic agent used in the denaturation of proteins and isolation of nucleic acids. This large quantity is ideal for high-throughput molecular biology labs that require significant volumes of denaturing reagent for large-scale protein analysis, RNA, and DNA extraction. Its denaturing properties are critical for disrupting protein structures, making it a standard reagent in studies of protein folding, enzyme function, and nucleic acid isolation. Researchers use this compound in protein chemistry, structural biology, and high-resolution assays where precise denaturation is essential for achieving reliable results.