Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanidine hydrochloride | 50-01-1 | 98.0% | 100 G
SDP

Catalog No. 5000407227
Encompass_Preferred

Guanidine hydrochloride | 50-01-1 | 98.0% | 100 G

Catalog No. 50-004-07227 Supplier Medchemexpress LLC Supplier No. HYB0178A100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guanidine hydrochloride | 50-01-1 | 98.0% | 100 G

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.