Learn More
Medchemexpress LLC Guanidine hydrochloride | 50-01-1 | 98.0% | 25 G
SDP
Catalog No. 5000406144
Encompass_Preferred
Guanidine hydrochloride is a strong chaotrope and denaturant of proteins, widely used in biochemical research. It causes co-operative unfolding of the molten globule state at high concentrations and can refold acid-unfolded proteins at low concentrations.
  • Induces co-operative unfolding of the molten globule state at concentrations greater than 1 M.
  • Refolds acid-unfolded apomyoglobin and cytochrome c at low concentrations.
  • Stabilizes the molten globule state.
  • Does not affect disulfide bonds.
  • Can cause efficient loss of the [PSI+] element from yeast cells at millimolar concentrations.
  • Reduces Hsp104-mediated basal and acquired thermotolerance.
  • Reduces Hsp104's ability to restore activity of thermally denatured luciferase.
  • In vivo, significantly reduces the death rate of infant mice infected with coxsackievirus A16.
Catalog No. 50-004-06144 Supplier Medchemexpress LLC Supplier No. 25GHYB0178A
Guanidine hydrochloride is a strong chaotrope and denaturant of proteins, widely used in biochemical research. It causes co-operative unfolding of the molten globule state at high concentrations and can refold acid-unfolded proteins at low concentrations.
  • Induces co-operative unfolding of the molten globule state at concentrations greater than 1 M.
  • Refolds acid-unfolded apomyoglobin and cytochrome c at low concentrations.
  • Stabilizes the molten globule state.
  • Does not affect disulfide bonds.
  • Can cause efficient loss of the [PSI+] element from yeast cells at millimolar concentrations.
  • Reduces Hsp104-mediated basal and acquired thermotolerance.
  • Reduces Hsp104's ability to restore activity of thermally denatured luciferase.
  • In vivo, significantly reduces the death rate of infant mice infected with coxsackievirus A16.