Learn More
Medchemexpress LLC Guanidine hydrochloride | 50-01-1 | 98.0% | 50 G
SDP
Catalog No. 5000406320
Encompass_Preferred
Guanidine hydrochloride is a strong chaotrope and a strong denaturant of proteins, intended for research use only.
  • Refolds acid-unfolded apomyoglobin and cytochrome c at low concentrations.
  • Causes co-operative unfolding of the molten globule state at concentrations greater than 1 M.
  • Does not affect ordered structure remaining due to disulfide bonds.
  • Causes efficient loss of the normally stable [PSI+] element from yeast cells at millimolar concentrations.
  • Reduces Hsp104-mediated basal and acquired thermotolerance.
  • Reduces Hsp104's ability to restore activity of thermally denatured luciferase.
  • Significantly reduced death rate of infant mice infected with coxsackievirus A16 in vivo.
Catalog No. 50-004-06320 Supplier Medchemexpress LLC Supplier No. HYB0178A50G
Guanidine hydrochloride is a strong chaotrope and a strong denaturant of proteins, intended for research use only.
  • Refolds acid-unfolded apomyoglobin and cytochrome c at low concentrations.
  • Causes co-operative unfolding of the molten globule state at concentrations greater than 1 M.
  • Does not affect ordered structure remaining due to disulfide bonds.
  • Causes efficient loss of the normally stable [PSI+] element from yeast cells at millimolar concentrations.
  • Reduces Hsp104-mediated basal and acquired thermotolerance.
  • Reduces Hsp104's ability to restore activity of thermally denatured luciferase.
  • Significantly reduced death rate of infant mice infected with coxsackievirus A16 in vivo.