Learn More
Alkali Scientific Guanidine Hydrochloride, 500 G
SDP
Catalog No. 5000659602
Encompass_Preferred
Guanidine Hydrochloride, available in a 500g format, is a powerful chaotropic agent used extensively in molecular biology for protein denaturation and nucleic acid isolation. It disrupts protein structures by breaking hydrogen bonds and other stabilizing interactions, making it ideal for extracting nucleic acids or for use in protein unfolding studies. This reagent is commonly used in protocols for RNA, DNA, and protein purification, especially in the preparation of samples for further biochemical analysis. Its versatility extends to use in protein-protein interaction studies and structural analysis of biomolecules in research labs.
Catalog No. 50-006-59602 Supplier Alkali Scientific Supplier No. GUA003CP
Guanidine Hydrochloride, available in a 500g format, is a powerful chaotropic agent used extensively in molecular biology for protein denaturation and nucleic acid isolation. It disrupts protein structures by breaking hydrogen bonds and other stabilizing interactions, making it ideal for extracting nucleic acids or for use in protein unfolding studies. This reagent is commonly used in protocols for RNA, DNA, and protein purification, especially in the preparation of samples for further biochemical analysis. Its versatility extends to use in protein-protein interaction studies and structural analysis of biomolecules in research labs.