Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Hydrochloride, BiotechGrade, 99%, Spectrum™ Chemical
SDP

Catalog No. 18603077 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-077 100 g Amber Glass Bottle
18-603-078 500 g Amber Glass Bottle
18-603-079 2.5 kg Amber Glass Bottle
18-603-081 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-077 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. GU103100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

GU103, 50-01-1, CH5N3·HCl

Spectrum™ Chemical Guanidine Hydrochloride, BiotechGrade is considered a chaotrope and a strong denaturants utilized in physiochemical studies of protein folding. Proteins become randomly coiled at high concentrations of guanidinium chloride. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
IUPAC Name hydrogen guanidine chloride
SMILES [H+].[Cl-].NC(N)=N

Specifications

CAS Max % 100%
Melting Range 184-188°C
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 99%
Grade Biotech
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Guanidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.