Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Guanidine Hydrochloride (GuHCl), 5.7 g, Solid, Reagent

Catalog No. 502391970 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Guanidine Hydrochloride (GuHCl) is a strong denaturant and chaotropic agent used for protein unfolding in digestions. It is supplied as a 5.7 g powder, enough for approximately 48 samples.

  • Strong denaturant and chaotropic agent for protein unfolding
  • 5.7 g powder in amber glass bottle
  • Sufficient for approximately 48 samples
  • Detection: MS, optical, UV
  • Formulation: solid
  • Storage: 2–8°C
  • System: LC/MS (HPLC, UPLC, UHPLC)
  • Shelf life: 24 months (6 months reconstituted)
  • Shipping: ambient
  • Application: reagent

Catalog No. 50-239-1970 Supplier Waters Corp Supplier No. 186010606
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guanidine Hydrochloride (GuHCl) is a strong denaturant and chaotropic agent used for protein unfolding in digestions. It is supplied as a 5.7 g powder, enough for approximately 48 samples.

  • Strong denaturant and chaotropic agent for protein unfolding
  • 5.7 g powder in amber glass bottle
  • Sufficient for approximately 48 samples
  • Detection: MS, optical, UV
  • Formulation: solid
  • Storage: 2–8°C
  • System: LC/MS (HPLC, UPLC, UHPLC)
  • Shelf life: 24 months (6 months reconstituted)
  • Shipping: ambient
  • Application: reagent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.