Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Hydrochloride, Practical Grade, MP Biomedicals

Catalog No. MP021019052 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021019052 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021019052 Supplier MP Biomedicals, Inc Supplier No. 02101905.2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Guanidine Hydrochloride is a strong chaotropic agent useful for the denaturation and subsequent refolding of proteins
  • This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies and be used for the recovery of periplasmic proteins
  • This can be used as the first step in refolding proteins or enzymes into their active form
  • Urea and dithiothreitol (DTT) may also be necessary
  • It is also used in the isolation of RNA to dissociate the nucleoprotein into its nucleic acid and protein moieties
  • It is an inhibitor of RNase

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 175°C to 190°C
Color White
Linear Formula CH5N3·HCl
Quantity 250 g
Solubility Information Soluble in (6M aq soln): Relatively Clear to Cloudy Yellow Solution with Sediment on Standing.
Formula Weight 95.53
Grade Practical
Physical Form Powder
Chemical Name or Material GUANIDINE HYDROCHLORIDE
Recommended Storage Store at Room Temperature (15-30°C). Store Desiccated.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.