Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Hydrochloride, Ultra Pure grade, MP Biomedicals

Catalog No. mp04820512 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04820512 100 g
MP04820540 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP04820512 Supplier MP Biomedicals, Inc Supplier No. 04820512
Only null left
Add to Cart
Add to Cart

  • Guanidine Hydrochloride is a strong chaotropic agent useful for the denaturation and subsequent refolding of proteins
  • This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies
  • This can be used as the first step in refolding proteins or enzymes into their active form
  • Urea and dithiothreitol (DTT) may also be necessary
  • Guanidine HCl is used in the isolation of RNA to dissociate the nucleoprotein into its nucleic acid and protein moieties
  • It is an inhibitor of RNase
  • Highly concentrated (6 - 8 M) Guanidine HCl solutions are used to denature native globular proteins
  • It apparently disrupts hydrogen bonds which hold the protein in its unique structure; however, there also is evidence suggesting that guanidine hydrocholoride may disrupt hydrophobic interactions by promoting the solubility of hydrophobic residues in aqueous solutions

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 180°C to 190°C
Color White
Linear Formula CH5N3·HCl
Quantity 100 g
Solubility Information Soluble in water (6 M - clear, colorless solution).
Formula Weight 95.53
Percent Purity ≥99%
Grade Ultra Pure
Physical Form Crystalline Powder
Chemical Name or Material GUANIDINE HYDROCHLORIDE
Recommended Storage Store at Room Temperature (15-30°C). Store Desiccated.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.