Learn More
Medchemexpress LLC Guanidine, N-(4-aminobutyl)-, sulfate | 2482-00-0 | MFCD00013109 | 98.0% | 228.27 | C5H16N4O4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10123810MM 1ML
Encompass_Preferred
Agmatine sulfate is a research reagent used in neuroscience and pharmacology to study imidazoline receptor signaling, nitric oxide synthesis, and neurotransmitter modulation. It is supplied in solid form and as a ready-to-use aqueous solution to support in vitro and biochemical experiments.
  • Acts as an endogenous agonist at imidazoline receptors.
  • Inhibits nitric oxide synthase and modulates NO pathways.
  • Modulates neurotransmitter systems and ion channels.
  • Available as solid material and as a ready-to-use 10 mM aqueous solution (1 mL) for convenience.
  • High purity suitable for research applications (reported ≈98.0%).
  • Molecular weight 228.27; CAS 2482-00-0.
Catalog No. 50-002-12446