Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanidine, N-methyl-, hydrochloride (1:1) | 21770-81-0 | 109.56 | 10 G
SDP

Catalog No. 5000445480
Encompass_Preferred

1-Methylguanidine monohydrochloride is an endogenous metabolite for research applications. This compound presents as a white to off-white solid with a high purity of 98.0%. It is suitable for various research fields including metabolic disease and as a reference standard.

  • Endogenous metabolite
  • High purity of 98.0%
  • Appears as a white to off-white solid
  • Store at 4°C, sealed, away from moisture
  • Stock solutions in solvent can be stored at -80°C for 6 months or -20°C for 1 month
  • Soluble in water and PBS

Catalog No. 50-004-45480 Supplier Medchemexpress LLC Supplier No. HYW00502810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methylguanidine monohydrochloride is an endogenous metabolite for research applications. This compound presents as a white to off-white solid with a high purity of 98.0%. It is suitable for various research fields including metabolic disease and as a reference standard.

  • Endogenous metabolite
  • High purity of 98.0%
  • Appears as a white to off-white solid
  • Store at 4°C, sealed, away from moisture
  • Stock solutions in solvent can be stored at -80°C for 6 months or -20°C for 1 month
  • Soluble in water and PBS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.