Learn More
Medchemexpress LLC Guanidine, N-methyl-, hydrochloride (1:1) | 21770-81-0 | 109.56 | 500 MG
SDP
Catalog No. 5000447434
Encompass_Preferred
1-Methylguanidine monohydrochloride is provided as a white to off-white solid. It has been tested and complies with the given specifications. This product is intended for research use only and has not been fully validated for medical applications.
  • White to off-white solid appearance
  • Tested and complies with specifications
  • Storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture
Catalog No. 50-004-47434 Supplier Medchemexpress LLC Supplier No. HYW005028500MG
1-Methylguanidine monohydrochloride is provided as a white to off-white solid. It has been tested and complies with the given specifications. This product is intended for research use only and has not been fully validated for medical applications.
  • White to off-white solid appearance
  • Tested and complies with specifications
  • Storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture