Learn More
Alkali Scientific Guanidine Thiocyanate, 1 Kg
SDP
Supplier:  Alkali Scientific UAN001CP
Encompass_Preferred
Guanidine Thiocyanate (1kg) is a chaotropic agent widely used in molecular biology for nucleic acid isolation and protein denaturation. Its ability to disrupt cellular structures by breaking down hydrogen bonds and other non-covalent interactions makes it essential in RNA and DNA extraction protocols, especially when isolating nucleic acids from biological samples. The compound also plays a role in inhibiting RNases during RNA extraction, ensuring high yields of intact RNA. It is a crucial reagent in labs performing large-scale nucleic acid work, including forensics, gene expression studies, and protein-RNA interaction research.
Catalog No. 50-006-61045