Learn More
Research Products International Corp Guanidine Thiocyanate, 1 Kilogram
SDP
Catalog No. 50213357
Encompass_Preferred
Protein denaturant used for purifying proteins and nucleic acids.
Toxicity: LD50
Catalog No. 50-213-357 Supplier Research Products International Corp Supplier No. G540001000.0
Protein denaturant used for purifying proteins and nucleic acids.
Toxicity: LD50