Learn More
Crescent Chemical Co Inc GUANIDINE THIOCYANATE
SDP
Catalog No. 50248403
Encompass_Preferred
Guanidine thiocyanate molecular biology grade DNase/RNase not detected. Suitable for the isolation of RNA. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 39577.02 CAS 593-84-0
Catalog No. 50-248-403 Supplier Crescent Chemical Co Inc Supplier No. SE39577.02
Guanidine thiocyanate molecular biology grade DNase/RNase not detected. Suitable for the isolation of RNA. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 39577.02 CAS 593-84-0