Learn More
Crescent Chemical Co Inc GUANIDINE THIOCYANATE
Supplier: Crescent Chemical Co Inc SE39577.01
Guanidine thiocyanate molecular biology grade DNase/RNase not detected. Suitable for the isolation of RNA. Size - 250G Storage Conditions - +15 °C TO +30 °C Catalog Number - 39577.01 CAS 593-84-0