Learn More
Medchemexpress LLC Guanidine thiocyanate | 593-84-0 | 99.9% | 100 G
SDP
Catalog No. 5000409984
Encompass_Preferred
Guanidine thiocyanate is a strong protein denaturant and potent inhibitor of nucleases. It acts as a nucleic acid protector during the extraction of DNA and RNA from cells and is a common component of buffers used for nucleic acid extraction.
  • Functions as a strong protein denaturant.
  • Acts as a potent inhibitor of nucleases.
  • Serves as a nucleic acid protector during DNA and RNA extraction.
  • Is a common component of buffers used for nucleic acid extraction.
Catalog No. 50-004-09984 Supplier Medchemexpress LLC Supplier No. HYD0841100G
Guanidine thiocyanate is a strong protein denaturant and potent inhibitor of nucleases. It acts as a nucleic acid protector during the extraction of DNA and RNA from cells and is a common component of buffers used for nucleic acid extraction.
  • Functions as a strong protein denaturant.
  • Acts as a potent inhibitor of nucleases.
  • Serves as a nucleic acid protector during DNA and RNA extraction.
  • Is a common component of buffers used for nucleic acid extraction.