Learn More
Selleck Chemical LLC Guanidine thiocyanate E4830-1G
SDP
Catalog No. 5000043295
Encompass_Preferred
Guanidine thiocyanate is a potent inhibitor of nucleases and a powerful denaturant of proteins It is used for RNA isolation due to its strong chaotropic properties which effectively denature RNase enzymes in samples ensuring RNA stability and purity during extraction processes Nucleic acid extraction buffers containing guanidine thiocyanate effectively deactivate high-titer polioviruses found in potentially infectious materials
Catalog No. 50-000-43295 Supplier Selleck Chemical LLC Supplier No. E48301G
Guanidine thiocyanate is a potent inhibitor of nucleases and a powerful denaturant of proteins It is used for RNA isolation due to its strong chaotropic properties which effectively denature RNase enzymes in samples ensuring RNA stability and purity during extraction processes Nucleic acid extraction buffers containing guanidine thiocyanate effectively deactivate high-titer polioviruses found in potentially infectious materials