Learn More
Alkali Scientific Guanidine Thiocyanate 6M Solution, 500 Milliliters
SDP
Supplier:  Alkali Scientific NID002CP
Encompass_Preferred
Guanidine Thiocyanate, 6M solution, is designed for high-efficiency nucleic acid isolation from biological samples. This reagent is commonly used in RNA and DNA extraction protocols, where it functions to denature proteins and disrupt cell membranes. The solution's high concentration makes it ideal for applications requiring robust protein removal and nucleic acid purification. Typically used in molecular biology and clinical research labs, it provides a powerful tool for isolating high-quality nucleic acids, free from contaminating proteins. This 500mL solution is ideal for laboratories needing a steady supply of reagent for frequent sample preparation, ensuring high yields in various nucleic acid-based experiments.
Catalog No. 50-006-61424