Learn More
Medchemexpress LLC Guanidinoacetic acid (13C2, 15N) | 2483829-93-0 | MFCD00004278 | 97.4% | 120.09 g/mol | C3H7N3O2 (contains 13C2, 15N) | 1 MG
SDP
Catalog No. 5000299510
Encompass_Preferred
Glycocyamine-15N,13C2 is an isotopically labeled form of guanidinoacetic acid used as an internal standard and tracer in metabolic and analytical studies. It contains two 13C labels and one 15N label, providing a defined mass shift for quantitative mass spectrometry and isotope tracing in small-sample workflows.
  • Isotopically labeled with two 13C and one 15N.
  • High isotopic enrichment (>99.1%).
  • Purity 97.4% (HPLC).
  • Suitable as an internal standard for LC-MS/MS and metabolic tracing.
  • Packaged in low-mass amounts for analytical use.
Catalog No. 50-002-99510 Supplier Medchemexpress LLC Supplier No. HYW021448S11MG
Glycocyamine-15N,13C2 is an isotopically labeled form of guanidinoacetic acid used as an internal standard and tracer in metabolic and analytical studies. It contains two 13C labels and one 15N label, providing a defined mass shift for quantitative mass spectrometry and isotope tracing in small-sample workflows.
  • Isotopically labeled with two 13C and one 15N.
  • High isotopic enrichment (>99.1%).
  • Purity 97.4% (HPLC).
  • Suitable as an internal standard for LC-MS/MS and metabolic tracing.
  • Packaged in low-mass amounts for analytical use.