Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanidinoacetic acid-d2 | 1173020-63-7 | 99.9% | 119.12 | C3H5D2N3O2 | 100 MG
SDP

Catalog No. 5000300492
Encompass_Preferred

Glycocyamine-d2 (guanidinoacetic acid-d2) is the deuterium-labeled form of glycocyamine used as a stable isotope reagent and internal standard for analytical assays and metabolic tracing. It is provided at high isotopic and chemical purity for accurate quantitation in LC-MS/MS and related methods.

  • Deuterium-labeled (d2) for reliable isotope tracing.
  • High purity (99.90%) suitable for quantitative analysis.
  • Molecular weight 119.12 and formula C3H5D2N3O2 for reference.
  • Available in small package sizes for analytical workflows.
  • Applicable as an internal standard in LC-MS/MS and metabolic studies.

Catalog No. 50-003-00492 Supplier Medchemexpress LLC Supplier No. HYW021448S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycocyamine-d2 (guanidinoacetic acid-d2) is the deuterium-labeled form of glycocyamine used as a stable isotope reagent and internal standard for analytical assays and metabolic tracing. It is provided at high isotopic and chemical purity for accurate quantitation in LC-MS/MS and related methods.

  • Deuterium-labeled (d2) for reliable isotope tracing.
  • High purity (99.90%) suitable for quantitative analysis.
  • Molecular weight 119.12 and formula C3H5D2N3O2 for reference.
  • Available in small package sizes for analytical workflows.
  • Applicable as an internal standard in LC-MS/MS and metabolic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.