Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanidinoacetic acid-d2 | 1173020-63-7 | 99.9% | 119.12 g/mol | C3H5D2N3O2 | 50 MG
SDP

Catalog No. 5000299511
Encompass_Preferred

Glycocyamine-d2 (guanidinoacetic acid-d2) is a deuterium-labeled isotopologue of glycocyamine supplied as a high-purity reference standard for analytical, tracer, and metabolic studies. It is intended for use in mass spectrometry, NMR, and metabolic pathway investigations where isotopic labeling improves quantitation and identification.

  • Deuterium-labeled (d2) isotopologue suitable for tracer and metabolism studies.
  • High purity (99.9%) for consistent analytical performance.
  • Molecular formula C3H5D2N3O2; molecular weight 119.12 g/mol.
  • Available in milligram pack sizes for standards and method development.
  • Applicable to mass spectrometry and NMR workflows for accurate quantitation.

Catalog No. 50-002-99511 Supplier Medchemexpress LLC Supplier No. HYW021448S50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Glycocyamine-d2 (guanidinoacetic acid-d2) is a deuterium-labeled isotopologue of glycocyamine supplied as a high-purity reference standard for analytical, tracer, and metabolic studies. It is intended for use in mass spectrometry, NMR, and metabolic pathway investigations where isotopic labeling improves quantitation and identification.

  • Deuterium-labeled (d2) isotopologue suitable for tracer and metabolism studies.
  • High purity (99.9%) for consistent analytical performance.
  • Molecular formula C3H5D2N3O2; molecular weight 119.12 g/mol.
  • Available in milligram pack sizes for standards and method development.
  • Applicable to mass spectrometry and NMR workflows for accurate quantitation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.