Learn More
TARGETMOL CHEMICALS INC Guanidinoethyl sulfonate 500MG
SDP
Catalog No. 502877467
Encompass_Preferred
Also available in 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Guanidinoethyl sulfonate (Taurocyamine) is a competitive glycine receptor antagonist.
Catalog No. 50-287-7467 Supplier TARGETMOL CHEMICALS INC Supplier No. T21615500MG
Also available in 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Guanidinoethyl sulfonate (Taurocyamine) is a competitive glycine receptor antagonist.