Learn More
Medchemexpress LLC Guanidinoethyl sulfonate | 543-18-0 | MFCD00084865 | 99.9% | 167.19 g/mol | C3H9N3O3S | 5 MG
SDP
Catalog No. 5000263306
Encompass_Preferred
Guanidinoethyl sulfonate (taurocyamine) is a small-molecule research reagent that inhibits taurine transport and antagonizes glycine receptors in the central nervous system. It is blood-brain barrier permeable and has been used in neuroprotection studies of ischemic brain injury; it also exhibits weak modulatory effects on GABAA receptors.
Catalog No. 50-002-63306 Supplier Medchemexpress LLC Supplier No. HY113329R5MG
Guanidinoethyl sulfonate (taurocyamine) is a small-molecule research reagent that inhibits taurine transport and antagonizes glycine receptors in the central nervous system. It is blood-brain barrier permeable and has been used in neuroprotection studies of ischemic brain injury; it also exhibits weak modulatory effects on GABAA receptors.