Learn More
Medchemexpress LLC Guanidinosuccinic acid | 6133-30-8 | 98.0% | 175.14 | 1 ML
SDP
Catalog No. 5000436091
Encompass_Preferred
Guanidinosuccinic acid is a nitrogenous metabolite that is elevated in the urine and serum of azotemic patients. It is a guanidino compound that has been shown to induce long-lasting generalized clonic and clonic-tonic convulsions in a dose-dependent manner in adult mice when administered intraperitoneally. In young mice, an age-dependent decrease in Guanidinosuccinic acid-induced convulsions and brain concentration was observed. The product is for research use only and not intended for sale to patients.
  • Endogenous metabolite
  • Elevated in azotemic patients
  • Exhibits epileptogenic and toxic properties in mice
Catalog No. 50-004-36091 Supplier Medchemexpress LLC Supplier No. HY11337310MM/1ML
Guanidinosuccinic acid is a nitrogenous metabolite that is elevated in the urine and serum of azotemic patients. It is a guanidino compound that has been shown to induce long-lasting generalized clonic and clonic-tonic convulsions in a dose-dependent manner in adult mice when administered intraperitoneally. In young mice, an age-dependent decrease in Guanidinosuccinic acid-induced convulsions and brain concentration was observed. The product is for research use only and not intended for sale to patients.
  • Endogenous metabolite
  • Elevated in azotemic patients
  • Exhibits epileptogenic and toxic properties in mice