Learn More
Medchemexpress LLC Guanidinosuccinic acid | 6133-30-8 | 98.0% | 175.14 | 100 MG
SDP
Catalog No. 5000436685
Encompass_Preferred
Guanidinosuccinic acid is a nitrogenous metabolite. It is a constituent of normal urine and is elevated in the urine and serum of azotemic patients. Guanidinosuccinic acid can induce long-lasting generalized clonic and clonic-tonic convulsions in adult mice in a dose-dependent manner.
  • Endogenous metabolite
  • Solid appearance, white to off-white color
  • For research use only
  • Shipped at room temperature
  • Sealed storage at 4°C, away from moisture
Catalog No. 50-004-36685 Supplier Medchemexpress LLC Supplier No. HY113373100MG
Add to Cart
Add to Cart
Guanidinosuccinic acid is a nitrogenous metabolite. It is a constituent of normal urine and is elevated in the urine and serum of azotemic patients. Guanidinosuccinic acid can induce long-lasting generalized clonic and clonic-tonic convulsions in adult mice in a dose-dependent manner.
  • Endogenous metabolite
  • Solid appearance, white to off-white color
  • For research use only
  • Shipped at room temperature
  • Sealed storage at 4°C, away from moisture