Learn More
Medchemexpress LLC Guanine-13C | 201489-20-5 | 99.5% | 152.12 g/mol | C413CH5N5O | 1mg
SDP
Catalog No. 5000268133
Encompass_Preferred
Guanine-13C is the 13C labeled Guanine[1] Guanine (2-Aminohypoxanthine) is one of the fundamental components of nucleic acids (DNA and RNA) Guanine is a purine derivative consisting of a fused pyrimidine-imidazole ring system with conjugated double bonds
Catalog No. 50-002-68133 Supplier Medchemexpress LLC Supplier No. HYY1055S1MG
Guanine-13C is the 13C labeled Guanine[1] Guanine (2-Aminohypoxanthine) is one of the fundamental components of nucleic acids (DNA and RNA) Guanine is a purine derivative consisting of a fused pyrimidine-imidazole ring system with conjugated double bonds