Learn More
Medchemexpress LLC Guanine-13C5 | 95.0% | 156.09 g/mol | 13C5H5N5O | 5mg
SDP
Catalog No. 5000267434
Encompass_Preferred
Guanine-13C5 is 13C-labeled Guanine (HY-Y1055) Guanine (2-Aminohypoxanthine) is one of the fundamental components of nucleic acids (DNA and RNA) Guanine is a purine derivative consisting of a fused pyrimidine-imidazole ring system with conjugated double bonds
Catalog No. 50-002-67434 Supplier Medchemexpress LLC Supplier No. HYY1055S45MG
Guanine-13C5 is 13C-labeled Guanine (HY-Y1055) Guanine (2-Aminohypoxanthine) is one of the fundamental components of nucleic acids (DNA and RNA) Guanine is a purine derivative consisting of a fused pyrimidine-imidazole ring system with conjugated double bonds