Learn More
Cayman Chemical Guanine-15N5, Purity: ≥98%, CAS: 168566-53-8, 5 mg
SDP
Catalog No. 502978363
Encompass_Preferred
An internal standard for the quantification of guanine by GC- or LC-MS
Catalog No. 50-297-8363 Supplier Cayman Chemical Supplier No. 411805 MG
An internal standard for the quantification of guanine by GC- or LC-MS