Learn More
Apexbio Technology LLC Guanine 73-40-5 5g
SDP
Catalog No. 502240588
Encompass_Preferred
Guanine (CAS 73-40-5) is a purine nucleobase characterized by a fused pyrimidine and imidazole ring system with conjugated double bonds As an integral component of DNA and RNA guanine forms hydrogen bonds with cytosine to facilitate nucleic acid base pairing thus playing a central role in the storage and transmission of genetic information It is frequently utilized in research investigating nucleic acid structure replication mutagenesis and enzymatic processes involving nucleotide metabolism or signaling pathways
This item is not returnable. View return policy
Catalog No. 50-224-0588 Supplier Apexbio Technology LLC Supplier No. B78895000
Guanine (CAS 73-40-5) is a purine nucleobase characterized by a fused pyrimidine and imidazole ring system with conjugated double bonds As an integral component of DNA and RNA guanine forms hydrogen bonds with cytosine to facilitate nucleic acid base pairing thus playing a central role in the storage and transmission of genetic information It is frequently utilized in research investigating nucleic acid structure replication mutagenesis and enzymatic processes involving nucleotide metabolism or signaling pathways