Learn More
Medchemexpress LLC Guanine | 73-40-5 | MFCD00071533 | 100.0% | 151.13 | C5H5N5O | 250mg
SDP
Catalog No. 5000268131
Encompass_Preferred
Guanine (Standard) is the analytical standard of Guanine This product is intended for research and analytical applications Guanine is one of the fundamental components of nucleic acids (DNA and RNA) Guanine is a purine derivative consisting of a fused pyrimidine-imidazole ring system with conjugated double bonds Guanine has the potential to serve as a large-capacity N pool[1]
Catalog No. 50-002-68131 Supplier Medchemexpress LLC Supplier No. HYY1055R250MG
Guanine (Standard) is the analytical standard of Guanine This product is intended for research and analytical applications Guanine is one of the fundamental components of nucleic acids (DNA and RNA) Guanine is a purine derivative consisting of a fused pyrimidine-imidazole ring system with conjugated double bonds Guanine has the potential to serve as a large-capacity N pool[1]