Learn More
Selleck Chemical LLC Guanine S4888-25mg
SDP
Catalog No. 503111396
Encompass_Preferred
Guanine is one of the four main nucleobases found in the nucleic acids DNA and RNA
Catalog No. 50-311-1396 Supplier Selleck Chemical LLC Supplier No. S488825MG
Guanine is one of the four main nucleobases found in the nucleic acids DNA and RNA